missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-53BP1 (S25) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF3405
This item is not returnable.
View return policy
Description
53BP1 Polyclonal specifically detects 53BP1 in Human samples. It is validated for Western Blot.
Specifications
| 53BP1 (p ser25) | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| TP53BP1 | |
| Phosphopeptide containing human 53BP1 S25 site | |
| 100 μg | |
| Primary | |
| Detects human 53BP1 when phosphorylated at S25. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, TP53BP1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7158 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction