missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PGRN ELISA Kit

Product Code. 17195139
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17195139

Brand: Invitrogen™ EH367RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human PGRN quantitates human PGRN in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PGRN.

Progranulin is the precursor protein of Granulin also called as Proepithelin and PC cell-derived growth factor. Cleavage of the signal peptide produces mature Granulin. Both Granulin and smaller cleavage product of Granulin regulates cell growth. Progranulin stimulates proliferation of many epithelial cells, promotes their anchorage-independent growth and, when overexpressed, confers epithelial tumorigenicity. Progranulin/epithelin gene is multifunctional, with important constitutive roles in epithelial homeostasis, reproductive, immunological, and neuronal function. Progranulin constitutively expressed in a number of epithelia, particularly in the skin, GI tract, and reproductive system. Progranulin is expressed in immune cells in vivo and in specific neurons in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Acrogranin; acrogranulin; CLN11; epithelin; Epithelin precursor; Epithelin/granulin precursor; Epithelin-1; Epithelin-2; GEP; Glycoprotein of 88 Kda; GP88; granulin; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; granulins; GRN; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; progranulin
Assay Range 0.1 to 25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2896
Gene Symbol GRN
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Progranulin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.