Learn More
Abnova™ Human PGBD1 Full-length ORF (NP_115896.1, 1 a.a. - 809 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_115896.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 84547 |
| Molecular Weight (g/mol) | 118.9kDa |
| Name | PGBD1 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MYEALPGPAPENEDGLVKVKEEDPTWEQVCNSQEGSSHTQEICRLRFRHFCYQEAHGPQEALAQLRELCHQWLRPEMHTKEQIMELLVLEQFLTILPKELQPCVKTYPLESGEEAVTVLENLETGSGDTGQQASVYIQGQDMHPMVAEYQGVSLECQSLQLLPGITTLKCEPPQRPQGNPQEVSGPVPHGSAHLQEKNPRDKAVVPVFNPVRSQTLVKTEEETAQAVAAEKWSHLSLTRRNLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSELLQGLSFSGDSDVEKDNEPEIQPAQKKLKVSCFPEKSWTKRDIKPNFPSWSALDSGLLNLKSEKLNPVELFELFFDDETFNLIVNETNNYASQKNVSLEVTVQEMRCVFGVLLLSGFMRHPRREMYWEVSDTDQNLVRDAIRRDRFELIFSNLHFADNGHLDQKDKFTKLRPLIKQMNKNFLLYAPLEEYYCFDKSMCECFDSDQFLNGKPIRIGYKIWCGTTTQGYLVWFEPYQEESTMKVDEDPDLGLGGNLVMNFADVLLERGQYPYHLCFDSFFTSVKLLSALKKKGVRATGTIRENRTEKCPLMNVEHMKKMKRGYFDFRIEENNEIILCRWYGDGIISLCSNAVGIEPVNEVSCCDADNEEIPQISQPSIVKVYDECKEGVAKMDQIISKYRVRIRSKKWYSILVSYMIDVAMNNAWQLHRACNPGASLDPLDFRRFVAHFYLEHNAHLSD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.