missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PFKFB4 (NP_004558) VersaClone cDNA
Description
- PRKRB4 is one of four bifunctional kinase/phosphatases that regulate the concentration of the glycolytic byproduct fructose-2,6- bisphosphate (F2,6BP).
- It is highly expressed in cancer cells and is induced by hypoxia.
- PRKRB4 is essential to the survival of cancer cells under conditions of hypoxia, because it increases the amount of F2,6BP and ATP at a time when the cell cannot produce much of them.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | PFKFB4 |
| Accession Number | NP_004558 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1423 bp |
| Gene ID (Entrez) | 5210 |
| Gene Alias | 6 PF-2-K/Fru-2,6-P2ase 4, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4,6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4,6 PF-2-K/Fru-2,6-P2ase 4,6 PF-2-K/Fru-2,6-P2ase testis-type isozyme, bifunctional enzyme with kinase and biphosphatase activities, PFK/FBPase 4 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?