missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Peroxiredoxin 6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£180.00 - £388.00
Specifications
| Antigen | Peroxiredoxin 6 |
|---|---|
| Clone | 477068R |
| Dilution | Western Blot 0.5 ug/mL, Knockout Validated |
| Applications | Western Blot, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705279
![]() |
R&D Systems
MAB3490R-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776659
![]() |
R&D Systems
MAB3490R-100 |
100 μg |
£388.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peroxiredoxin 6 Monoclonal specifically detects Peroxiredoxin 6 in Human samples. It is validated for Western Blot.Specifications
| Peroxiredoxin 6 | |
| Western Blot 0.5 ug/mL, Knockout Validated | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P30041 | |
| 9588 | |
| IgG3 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 477068R | |
| Western Blot, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 1-Cys, 24 kDa protein, Acidic calcium-independent phospholipase A2, aiPLA2, aiPLA21-Cys PRX, Antioxidant protein 2, AOP2, AOP2Non-selenium glutathione peroxidase, EC 1.11.1.15, EC 1.11.1.7, EC 3.1.1.-, KIAA0106Red blood cells page spot 12, Liver 2D page spot 40, MGC46173, NSGPx, NSGPx1-Cys peroxiredoxin, p29, peroxiredoxin 6, peroxiredoxin-6, PRDX6, PRX | |
| E. coli-derived recombinant human Peroxiredoxin 6 Met1-Pro224 Accession # P30041 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Peroxiredoxin 6 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title