missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Perilipin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB76341-SP
This item is not returnable.
View return policy
Description
Perilipin-2/ADFP Monoclonal specifically detects Perilipin-2/ADFP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Perilipin-2/ADFP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ADFP, Adipophilin, Adipose differentiation-related proteinadipophilin, ADRP, MGC10598, perilipin 2, perilipin-2, Perilipin2, PLIN2 | |
| E. coli-derived recombinant human Perilipin-2 Met1-His437 Accession # Q99541 | |
| 25 μg | |
| 123 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 788616 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q99541 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Perilipin-2 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction