missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Perforin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£177.00 - £428.00
Specifications
| Antigen | Perforin |
|---|---|
| Clone | 1001103 |
| Dilution | Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793627
![]() |
R&D Systems
MAB8011-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18781367
![]() |
R&D Systems
MAB8011-100 |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Perforin Monoclonal specifically detects Perforin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Perforin | |
| Western Blot 2 μg/mL, Immunohistochemistry 5-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1 | |
| Synthetic peptide containing human Perforin, Pro22-Trp555, Accession No. P14222 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Perforin in direct ELISAs. |
| 1001103 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human | |
| P14222 | |
| 5551.0 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title