missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pentraxin 2/SAP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1948
This item is not returnable.
View return policy
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Western Blot.
Specifications
| Pentraxin 2/SAP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P02743 | |
| APCS | |
| Mouse myeloma cell line NS0-derived recombinant human Pentraxin 2/SAP His20-Val233 Accession # P02743 | |
| 100 μg | |
| Primary | |
| Detects human Pentraxin 2/SAP in ELISA and Western Blot. | |
| Human | |
| Purified |
| Western Blot | |
| 910119 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 325 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction