missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEI-Transferrinfection Kit
Click to view available options
Quantity:
16 Transferrinfections
32 Transferrinfections
Unit Size:
16 reactions
32 reactions
Description
Includes
- Transferrin-Polyethylenimine-conjugate, lyophilized, 1 mg/mL upon reconstitution; iron incorporated
- Deferrioxamine, lyophilied, 50 mM upon reconstitution
- HBS buffer concentrate (20x)
- (400mM Hepes, pH 7.3, 3M NaCl)
- Method results in a 30 to 1000-fold enhanced transfection efficiency depending on the cell line
- An extremely gentle DNA transfection method which employs physiological uptake mechanisms of the cell
- Transfection efficacy depends on the cell type, the level of surface transferrin receptor expression
- Very high transfection rates have been shown for the tested tumor cell lines B16F10 melanoma, Neuro 2A neuroblastoma and a variety of primary human melanoma cell lines
- In other established cell lines, such as HeLa, CHO, Jurkat, K562, HepG2 and COS the PEI-Transferrinfection works with high efficiences, excellent reproducibility and with the advantage of being an extremely gentle procedure
Specifications
Specifications
| Classification | Chemically Defined |
| Cell Type | Tumor Cell Lines, Established Cell Lines |
| Format | Kit |
| Sample Type | Plasmid DNA |
| Transfection Technique | Transferrin Receptor-mediated Endocytosis |
| For Use With (Application) | Transfection |
| Product Type | Transferrinfection Kit |
| Quantity | 16 Transferrinfections |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction