missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PEI-Transferrinfection Kit

Product Code. 15541307
Click to view available options
Quantity:
16 Transferrinfections
32 Transferrinfections
Unit Size:
16 reactions
32 reactions
This item is not returnable. View return policy

Product Code. 15541307

Brand: Invitrogen™ BMS1003a

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Transferrin-Polyethylenimine-conjugate, lyophilized, 1 mg/mL upon reconstitution; iron incorporated
  • Deferrioxamine, lyophilied, 50 mM upon reconstitution
  • HBS buffer concentrate (20x)
  • (400mM Hepes, pH 7.3, 3M NaCl)

Nucleic acid delivery system based on transferrin receptor-mediated endocytosis to carry DNA into cells and the gene transfer efficiency of PEI/DNA complexes

  • Method results in a 30 to 1000-fold enhanced transfection efficiency depending on the cell line
  • An extremely gentle DNA transfection method which employs physiological uptake mechanisms of the cell
  • Transfection efficacy depends on the cell type, the level of surface transferrin receptor expression
  • Very high transfection rates have been shown for the tested tumor cell lines B16F10 melanoma, Neuro 2A neuroblastoma and a variety of primary human melanoma cell lines
  • In other established cell lines, such as HeLa, CHO, Jurkat, K562, HepG2 and COS the PEI-Transferrinfection works with high efficiences, excellent reproducibility and with the advantage of being an extremely gentle procedure
TRUSTED_SUSTAINABILITY

Specifications

Classification Chemically Defined
Cell Type Tumor Cell Lines, Established Cell Lines
Format Kit
Sample Type Plasmid DNA
Transfection Technique Transferrin Receptor-mediated Endocytosis
For Use With (Application) Transfection
Product Type Transferrinfection Kit
Quantity 16 Transferrinfections
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.