missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PEG10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB99751-SP
This item is not returnable.
View return policy
Description
PEG10 Monoclonal specifically detects PEG10 in Human samples. It is validated for ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| PEG10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EDR, embryonal carcinoma differentiation regulated, Embryonal carcinoma differentiation-regulated protein, HB-1, KIAA1051EDR, Mammalian retrotransposon-derived protein 2, Mar2, MART2, MEF3 like 1, MEF3L, MEF3L1, Myelin expression factor 3-like protein 1, paternally expressed 10, Paternally expressed gene 10 protein, retrotransposon gag domain containing 3, Retrotransposon gag domain-containing protein 3, Retrotransposon-derived gag-like polyprotein, retrotransposon-derived protein PEG10, RGAG3, RGAG3MEF3-like protein 1, RTL2, SIRH1, Ty3/Gypsy-like protein | |
| Synthetic peptide containing human PEG10, Accession No. Q86TG7 | |
| 25 μg | |
| 23089.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA, Flow Cytometry, CyTOF | |
| 981902 | |
| ELISA, Intracellular Staining by Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
| Q86TG7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human PEG10 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction