missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PEDFR/PNPLA2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5387
This item is not returnable.
View return policy
Description
PEDFR/PNPLA2/ATGL Polyclonal specifically detects PEDFR/PNPLA2/ATGL in Human samples. It is validated for Immunohistochemistry.
Specifications
| PEDFR/PNPLA2/ATGL | |
| Polyclonal | |
| Unconjugated | |
| Q96AD5 | |
| PNPLA2 | |
| E. coli-derived recombinant human PEDF R/PNPLA2 Val162-Thr332 Accession # Q96AD5 | |
| 100 μg | |
| Primary | |
| Detects human PEDF R/PNPLA2 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse PNPLA2 is observed. | |
| Human | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| Adipose triglyceride lipase, ATGL, ATGL1110001C14Rik, Calcium-independent phospholipase A2, Desnutrin, EC 3.1.1.3, FP17548, IPLA2-zeta, patatin-like phospholipase domain containing 2, patatin-like phospholipase domain-containing protein 2, patatin-like phospholipase domain-containing protein 2-like, PEDF R, PEDF-R, Pigment epithelium-derived factor, PNPLA2, Transport-secretion protein 2, transport-secretion protein 2.2, triglyceride hydrolase, TTS2, TTS2.2, TTS-2.2, TTS2DKFZp667M109 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57104 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction