missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PEAR1 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human PEAR1 quantitates human PEAR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PEAR1.
Platelet endothelial aggregation receptor 1 (PEAR1) is a platelet receptor that signals upon the formation of platelet-platelet contacts independent of platelet activation and secondary to platelet aggregation. Upon platelet aggregation stimulated by physiological agonists, PEAR1 becomes tyrosine- and serine-phosphorylated; the tyrosine phosphorylation can be inhibited by eptifibatide, an aIIbbeta3 antagonist that also inhibits platelet aggregation. Recent studies have indicated that genetic variations in PEAR1, may be associated with enhanced agonist-induced platelet aggregation.
Specifications
Specifications
| Accession Number | 3110045G13Rik; hPEAR1; Jagged and Delta protein; Jedi; Jedi-1; Megf12; mPEAR1; multiple EGF-like domains protein 12; multiple EGF-like-domains 12; multiple epidermal growth factor-like domains protein 12; Pear1; platelet endothelial aggregation receptor 1; Protein Jedi; RGD1305653; RP11-107D16.3 |
| Assay Range | 0.45 to 100 ng/mL |
| Assay Sensitivity | 0.45 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 375033 |
| Gene Symbol | PEAR1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction