missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PEAR1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4527G-100UG
This item is not returnable.
View return policy
Description
PEAR1 Monoclonal specifically detects PEAR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| PEAR1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| JEDI, platelet endothelial aggregation receptor 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human PEAR1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Tenascin R or recombinant mouse Tenascin C is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 492621 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q5VY43 | |
| PEAR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human PEAR1, Leu21-Ser754, Accession # Q5VY43 | |
| 100 μg | |
| 375033 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction