missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PDSS1 (aa 319-412) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58217 (PA5-58217. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
PDSS1 is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. PDSS1 catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in PDSS1 gene are a cause of coenzyme Q10 deficiency.The protein encoded by this gene is an enzyme that elongates the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. The protein may be peripherally associated with the inner mitochondrial membrane, though no transit peptide has been definitively identified to date. Defects in this gene are a cause of coenzyme Q10 deficiency.
Specifications
Specifications
| Accession Number | Q5T2R2 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23590 |
| Name | Human PDSS1 (aa 319-412) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2610203G20Rik; 2700031G06Rik; All-trans-decaprenyl-diphosphate synthase subunit 1; COenzyme Q (ubiquinone) biosynthesis family member (coq-1)-like; coenzyme Q1 homolog; COQ1; COQ10D2; decaprenyl diphdsphate synthase subunit1; decaprenyl diphosphate synthase subunit 1; decaprenyl pyrophosphate synthase subunit 1; decaprenyl pyrophosphate synthetase subunit 1; decaprenyl-diphosphate synthase subunit 1; DPS; Dps1; hDPS1; hypothetical protein LOC550349; LOW QUALITY PROTEIN: decaprenyl-diphosphate synthase subunit 1; mDLP1; MGC70953; mSPS1; PDSS1; polyprenyl pyrophosphate synthetase; prenyl (decaprenyl) diphosphate synthase, subunit 1; prenyl (solanesyl) diphosphate synthase, subunit 1; prenyl diphosphate synthase, subunit 1; RP13-16H11.3; solanesyl-diphosphate synthase subunit 1; SPS; Sps1; subunit 1 of decaprenyl diphosphate synthase; subunit 1 of solanesyl diphosphate synthase; TPRT; TPT; TPT 1; trans-prenyltransferase; trans-prenyltransferase (TPT); trans-prenyltransferase 1; wu:fc11f12; wu:fd05d05; zgc:112058 |
| Common Name | PDSS1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction