missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF R beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB385
This item is not returnable.
View return policy
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Western Blot.
Specifications
| PDGF R beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 5159 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 61520 | |
| Western Blot 1 ug/mL | |
| P09619 | |
| PDGFRB | |
| Mouse myeloma cell line NS0-derived recombinant human PDGF R beta Leu33-Lys531 (Lys531Glu) Accession # P09619 | |
| 500 μg | |
| Primary | |
| Detects human PDGF R beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) PDGF R beta , rhPDGF R alpha , rhgp130, or rhIL-6 R. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction