missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF R beta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 24 publications
Brand: R&D Systems AF385-SP
This item is not returnable.
View return policy
Description
PDGF R beta Polyclonal specifically detects PDGF R beta in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Specifications
| PDGF R beta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5159 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 10-40 ug/mL, CyTOF-ready | |
| P09619 | |
| PDGFRB | |
| Mouse myeloma cell line NS0-derived recombinant human PDGF R beta Leu33-Phe530 (Glu241Asp) Accession # P09619 | |
| 25 μg | |
| Primary | |
| Detects human PDGF R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse PDGF R beta is observed, and less than 1% cross-reactivity with recombinant human PDGF sR alpha is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction