missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF R alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
£184.00 - £1775.00
Specifications
| Antigen | PDGFR alpha |
|---|---|
| Clone | 35248 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 2-10 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777768
![]() |
R&D Systems
MAB322-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742569
![]() |
R&D Systems
MAB322-100 |
100 μg |
£457.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18783307
![]() |
R&D Systems
MAB322-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PDGFR alpha Monoclonal specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| PDGFR alpha | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 2-10 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| PDGFRA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 35248 | |
| Western Blot, Immunohistochemistry, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5156 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PDGF R alpha | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PDGF R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human PDGF R beta or recombinant mouse PDGF R alpha is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title