missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PDGF-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB220-SP
This item is not returnable.
View return policy
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.
Specifications
| PDGF-B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5155 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 108132 | |
| Western Blot 1 ug/mL | |
| P01127 | |
| PDGFB | |
| E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 | |
| 25 μg | |
| Primary | |
| Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs, shows approximately 40% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhCTGF, rhLDGF, rhPDGF-AA, recombinant rat (rr) PDGF-AA, rrPDGF-AB, rrPDGF-BB, rhPDGF-C, rhPDGF-D, rhPlGF, rhVEGF, rhVEGF-C, or rhVEGF-D. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction