missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human PDGF-AB Quantikine ELISA Kit

Product Code. 18771834
Change view
Click to view available options
Sufficient For:
1 x 96-well plate
6 x 96-well plates
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18771834 1 x 96-well plate 1 set
18722264 6 x 96-well plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18771834 Supplier R&D Systems™ Supplier No. DHD00C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human PDGF-AB Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human PDGF-AB Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PDGF-AB in cell culture supernates (100 ul), serum (10 ul), platelet-poor edta plasma (100 ul), platelet-poor heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 3.83pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum
Specificity Natural and Recombinant Human PDGF-AB
Sufficient For 1 x 96-well plate
Target Species Human
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Serum (10uL),Platelet-poor EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.