missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6 gamma Polyclonal Antibody

Human PDE6 gamma (aa 1-15) Polyclonal Antibody
Brand: Thermo Scientific PA5106261
This item is not returnable.
View return policy
Description
Human PDE6 gamma (aa 1-15) Polyclonal Antibody for Western Blot
Specifications
| PDE6 gamma | |
| Polyclonal | |
| Unconjugated | |
| PDE6G | |
| GMP-PDE gamma; p3AT; PDE6G; PDEG; phosphodiesterase 6G; phosphodiesterase 6G, cGMP-specific, rod, gamma; phosphodiesterase, cyclic GMP (rod receptor), gamma polypeptide; retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma; rod cG-PDE G; RP57; unnamed protein product | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18588, 5148, 688297 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09174, P18545 | |
| PDE6G | |
| A synthesized peptide derived from human PDE6G(Accession P18545), corresponding to amino acid residues M1-F50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction