missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-L2/B7-DC Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB1224-100
This item is not returnable.
View return policy
Description
PD-L2/B7-DC/PDCD1LG2 Monoclonal specifically detects PD-L2/B7-DC/PDCD1LG2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| PD-L2/B7-DC/PDCD1LG2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 0.5-25 ug/mL | |
| Q9BQ51 | |
| PDCD1LG2 | |
| Mouse myeloma cell line NS0-derived recombinant human PD-L2/B7-DC Leu20-Pro219 (predicted) Accession # Q9BQ51 | |
| 100 μg | |
| Primary | |
| Detects human PD-L2/B7-DC in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse PD-L2, recombinant human (rh) B7-1, rhB7-2, rhB7-H1, rhB7-H2, or rhB7-H3 is observed. | |
| Human | |
| Supernatant |
| Western Blot, Immunohistochemistry, Immunohistochemistry, CyTOF | |
| 176611 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, Butyrophilin-like Protein, CD273, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDCD1LG2, PDL2, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 80380 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction