missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PD-1 His-tag Recombinant Protein

Product Code. p-7230008
Change view
Click to view available options
Quantity:
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
50µg
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17164203 50 μg 50µg
17154203 5 x 50 μg Pack of 5
17144203 5 x 5 μg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17164203 Supplier Invitrogen™ Supplier No. A42533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of 'danger signals' such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15116
For Use With (Application) Control, Binding Assay, Immunohistochemistry, Surface Plasmon Resonance, Western Blot
Formulation 50mM MOPS with 0.01% Tween 80, 5% mannitol, 500mM NaCl, 5% trehalose and no preservative; pH 7
Gene ID (Entrez) 5133
Molecular Weight (g/mol) 17.4 kDa
Name Human PD-1 His-tag
pH Range 7
Purification Method Purified
Quantity 50 μg
Source HEK293 cells
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
Common Name PD-1 (CD279)
Gene Symbol Pdcd1
Biological Activity 1. Immobilized Human PD-1 His-tag Recombinant Protein at 2 μg/mL (100 μL/well) can bind PD-L1 Protein, Human, Recombinant (ECD, hFc Tag), the EC50 of human B7-H1 is 150-600 ng/mL 2. Immobilized Human PD-1 His-tag Recombinant Protein (at 2 μg/mL (100 μL/well) can bind Anti-PD1 (MDX)-IgG4 Antibody (Nivolumab), the EC50 is 8-60 ng/mL 3. Loaded Anti-human PD1 antibody, IgG4 on ProA Biosensor, can bind Human PD-1 His-tag Recombinant Protein with an affinity constant of 2.44 nM as determined in a BLI assay (Sartorius Octet RED384) (Routinely tested). 4.Captured Anti-PD1 Mab (Human IgG4) on proA Chip can bind Human PD-1 His-tag Recombinant Protein with an affinity constant of 10.03 nM as determined in a SPR assay (Biacore T200) (Routinely tested).)
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Human PD1, amino acids Met1-Gln167 (Accession # NP_005009.2) with a C-terminal His-tag
Content And Storage Includes 1 vial of lyophilized protein. Store at -20 °C.
Activity 1. Immobilized Human PD-1 His-tag Recombinant Protein at 2 μg/mL (100 μL/well) can bind PD-L1 Protein, Human, Recombinant (ECD, hFc Tag), the EC50 of human B7-H1 is 150-600 ng/mL.<br><br>2. Immobilized Human PD-1 His-tag Recombinant Protein at 2 μg/mL (100 μL/well) can bind Anti-PD1 (MDX)-IgG4 Antibody (Nivolumab), the EC50 is 8-60 ng/mL.<br><br>3. Loaded Anti-human PD1 antibody, IgG4 on ProA Biosensor, can bind Human PD-1 His-tag Recombinant Protein with an affinity constant of 2.44 nM as determined in a BLI assay (Sartorius Octet RED384) (Routinely tested).<br><br>4.Captured Anti-PD1 Mab (Human IgG4) on proA Chip can bind Human PD-1 His-tag Recombinant Protein with an affinity constant of 10.03 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Endotoxin Level < 1 EU/μg
Expression System HEK293 cells
Protein Family CD Proteins
Form Lyophilized
Protein Subtype Other Proteins
Research Category Apoptosis, Immunology, Inflammation
Purity or Quality Grade >95 % as determined by SDS-PAGE. >90 % as determined by SEC-HPLC.
Protein programmed cell death 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.