missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PCID1 (aa 36-178) Control Fragment Recombinant Protein

Product Code. 30197356
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197356

Brand: Invitrogen™ RP101061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56626 (PA5-56626. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L2H7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10480
Name Human PCID1 (aa 36-178) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias B5; B5 receptor; dendritic cell protein; dendritic cell protein GA17; eIF3m; Eukaryotic translation initiation factor 3 subunit M; eukaryotic translation initiation factor 3, subunit M; fetal lung protein B5; FLJ29030; GA17; HFLB5; hFL-B5; PCI domain containing 1 (herpesvirus entry mediator); PCI domain-containing protein 1; PCID1; PNAS-125; RGD1565840; Tango7; transport and golgi organization 7 homolog
Common Name PCID1
Gene Symbol EIF3M
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GGLHVDLAQIIEACDVCLKEDDKDVESVMNSVVSLLLILEPDKQEALIESLCEKLVKFREGERPSLRLQLLSNLFHGMDKNTPVRYTVYCSLIKVAASCGAIQYIPTELDQVRKWISDWNLTTEKKHTLLRLLYEALVDCKKS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.