missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCGF1 Partial ORF (NP_116062, 105 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084759-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100887

  • £425.00 / 25µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PCGF1 is a mammalian homolog of the Drosophila polycomb group genes, which act as transcriptional repressors to regulate anterior-posterior patterning in early embryonic development (Nunes et al., 2001 [PubMed 11287196]). See also PCGF2 (MIM 600346).[supplied by OMIM]

Sequence: DSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAE
Specifications

Specifications

NP_116062
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.87kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAE
RUO
PCGF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84759
PCGF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2010002K04Rik/FLJ43754/MGC10882/NSPC1/RNF3A-2/RNF68
PCGF1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers