missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PBX2 (aa 364-405) Control Fragment Recombinant Protein

Product Code. 30206002
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30206002

Brand: Invitrogen™ RP100032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63964 (PA5-63964. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40425
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5089
Name Human PBX2 (aa 364-405) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU043397; G17; homeobox 12; homeobox protein PB x 2; HO x 12; PBX homeobox 2; Pb x 2; PB x 2 MHC; PB x 2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13
Common Name PBX2
Gene Symbol PBX2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GDSYSASQVESLRHSMGPGGYGDNLGGGQMYSPREMRANGSW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.