missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PBEF/Visfatin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£205.00 - £415.00
Specifications
| Antigen | PBEF/Visfatin/NAMPT |
|---|---|
| Clone | 882115R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18119270
![]() |
R&D Systems
MAB4335-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18146161
|
R&D Systems
MAB4335-100 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PBEF/Visfatin/NAMPT Monoclonal specifically detects PBEF/Visfatin/NAMPT in Human samples. It is validated for ELISA.Specifications
| PBEF/Visfatin/NAMPT | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P43490 | |
| 10135 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 882115R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 2.4.2.12, MGC117256, NAmPRTase, NAMPT, nicotinamide phosphoribosyltransferase, NMPRTase, PBEF1, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
| E. coli-derived recombinant Human PBEF/Visfatin Pro27-His491 Accession # P43490 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human PBEF/Visfatin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title