missing translation for 'onlineSavingsMsg'
Learn More

Human Pax3 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£200.00 - £491.00

Specifications

Antigen Pax3
Clone 274206
Dilution Immunohistochemistry 5-15 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18106081
View Documents
R&D Systems
MAB11430-SP
25 μg
£200.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18151471
GREENER_CHOICE
View Documents
R&D Systems
MAB11430-100
100 μg
£491.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pax3 Monoclonal antibody specifically detects Pax3 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

Pax3
Immunohistochemistry 5-15 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
E. coli-derived recombinant human PAX-3, Met1-Leu215, Accession # P23760
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
274206
Immunohistochemistry
Unconjugated
Mouse
Human
P23760
5077
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.