missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PATJ (aa 1597-1679) Control Fragment Recombinant Protein

Product Code. 30212737
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212737

Brand: Invitrogen™ RP103978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64275 (PA5-64275. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein with multiple PDZ domains. PDZ domains mediate protein-protein interactions, and proteins with multiple PDZ domains often organize multimeric complexes at the plasma membrane. This protein localizes to tight junctions and to the apical membrane of epithelial cells. A similar protein in Drosophila is a scaffolding protein which tethers several members of a multimeric signaling complex in photoreceptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NI35
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10207
Name Human PATJ (aa 1597-1679) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias channel-interacting PDZ domain protein; channel-interacting PDZ domain-containing protein; Cipp; hINADL; inactivation no after-potential D-like protein; inactivation-no-afterpotential D-like; Inadl; Inadl protein; Inadl2; InaD-like; InaD-like (Drosophila); InaD-like 2; inaD-like protein; novel ankyrin-repeat protein; PALS1-associated tight junction protein; Patj; PATJ, crumbs cell polarity complex component; PDZ domain protein; protein associated to tight junctions; RGD1565362
Common Name PATJ
Gene Symbol PATJ
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ATILKCAQGLVQLEIGRLRAGSWTSARTTSQNSQGSQQSAHSSCHPSFAPVITGLQNLVGTKRVSDPSQKNSGTDMEPRTVEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.