missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PARS2 (aa 340-431) Control Fragment Recombinant Protein

Product Code. 30201905
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201905

Brand: Invitrogen™ RP94379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55888 (PA5-55888. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a putative member of the class II family of aminoacyl-tRNA synthetases. These enzymes play a critical role in protein biosynthesis by charging tRNAs with their cognate amino acids. This protein is encoded by the nuclear genome but is likely to be imported to the mitochondrion where it is thought to catalyze the ligation of proline to tRNA molecules. Mutations have been found in this gene in some patients with Alpers syndrome. [provided by RefSeq, Mar 2015]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L3T8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25973
Name Human PARS2 (aa 340-431) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC027073; class II tRNA synthetase; MT-PRORS; PARS2; Probable proline--tRNA ligase, mitochondrial; probable prolyl-tRNA synthetase, mitochondrial; proline tRNA ligase 2, mitochondrial (putative); proline--tRNA ligase; Prolyl-tRNA synthetase; prolyl-tRNA synthetase (mitochondrial)(putative); prolyl-tRNA synthetase 2, mitochondrial (putative); proRS; RGD1305345
Common Name PARS2
Gene Symbol PARS2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YGLGVTRILAAAIEVLSTEDCVRWPSLLAPYQACLIPPKKGSKEQAASELIGQLYDHITEAVPQLHGEVLLDDRTHLTIGNRLKDANKFGYP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.