missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PARP6 (aa 497-579) Control Fragment Recombinant Protein

Product Code. 30201827
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201827

Brand: Invitrogen™ RP90670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55475 (PA5-55475. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Poly(ADP-ribosyl)ation is an immediate DNA-damage-dependent post-translational modification of histones and other nuclear proteins that contributes to the survival of injured proliferating cells. Poly(ADP-ribose) polymerases (PARPs) now constitute a large family of 18 proteins, encoded by different genes and displaying a conserved catalytic domain in which PARP-1 (113 kDa), the founding member, and PARP-2 (62 kDa) are so far the sole enzymes whose catalytic activity has been shown to be immediately stimulated by DNA strand breaks. A large repertoire of sequences encoding novel PARPs now extends considerably the field of poly(ADP-ribosyl)ation reactions to various aspects of the cell biology including cell proliferation and cell death. Some of these new members interact with each other, share common partners and common subcellular localizations suggesting possible fine tuning in the regulation of this post-translational modification of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2NL67
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56965
Name Human PARP6 (aa 497-579) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700119G14Rik; 2310028P13Rik; 3110038K10Rik; ADP-ribosyltransferase diphtheria toxin-like 17; ARTD17; C030013N01Rik; PARP6; PARP-6; PARP-6-B1; PARP-6-C; pART17; poly (ADP-ribose) polymerase family, member 6; poly [ADP-ribose] polymerase 6; poly(ADP-ribose) polymerase family member 6; Protein mono-ADP-ribosyltransferase PARP6; RGD1310937
Common Name PARP6
Gene Symbol Parp6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QLHGAAYGKGIYLSPISSISFGYSGMGKGQHRMPSKDELVQRYNRMNTIPQTRSIQSRFLQSRNLNCIALCEVITSKDLQKHG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.