missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PARL Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
PARL Synthetic Peptide, PEP-0699, from Invitrogen.
This gene encodes a mitochondrial integral membrane protein. Following proteolytic processing of this protein, a small peptide is formed and translocated to the nucleus. This gene may be involved in signal transduction via regulated intramembrane proteolysis of membrane-tethered precursor proteins. Variation in this gene has been associated with increased risk for type 2 diabetes. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Specifications
| Gene ID (Entrez) | 55486 |
| Species | Human |
| Purification Method | purified |
| Accession Number | Q9H300 |
| Common Name | Human PARL |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction