missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Park7/DJ-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3995-SP
This item is not returnable.
View return policy
Description
Park7/DJ-1 Polyclonal specifically detects Park7/DJ-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Knockout Validated.
Specifications
| Park7/DJ-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DJ1, DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11315 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
| Q99497 | |
| PARK7 | |
| E. coli-derived recombinant human Park7/DJ-1 Met1-Asp189 Accession # Q99497 | |
| 25 μg | |
| Primary | |
| Detects human Park7/DJ-1 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction