missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Parechovirus 3 VP3 (Strain: WA17c-06) Recombinant Rabbit Monoclonal Antibody (HL2544)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556754
This item is not returnable.
View return policy
Description
Human Parechovirus 3 VP3 (Strain: WA17c-06) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The Human Parechovirus Type 3 (HPeV-3) VP3 protein is a vital structural component of the HPeV-3 virus, which belongs to the Picornaviridae family. The VP3 protein is one of the capsid proteins that form the outer shell of the virus, encapsulating and protecting its RNA genome. This protein plays a crucial role in the virus's ability to infect host cells, as it is involved in the initial attachment and entry into the host cell, as well as in the assembly and stability of the viral particle. HPeV-3 is associated with a range of clinical manifestations, particularly severe in infants and young children, including neonatal sepsis, meningitis, encephalitis, and respiratory infections. The VP3 protein is highly immunogenic, making it an important target for the development of diagnostic tools, vaccines, and antiviral therapies. Understanding the structure and function of the VP3 protein in HPeV-3 is essential for advancing research into the pathogenic mechanisms of the virus and for creating effective strategies to prevent and treat HPeV-3 infections.
Specifications
| Human Parechovirus 3 VP3 (Strain: WA17c-06) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL2544 | |
| Unconjugated | |
| Human PeV 3 VP3 | |
| Synthetic peptide of Human Parechovirus 3 VP3. (strain: WA17c-06). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction