missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Parechovirus 1/3 VP3 (Strain: WA17c-06) Recombinant Rabbit Monoclonal Antibody (HL2443)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556753
This item is not returnable.
View return policy
Description
Human Parechovirus 1/3 VP3 (Strain: WA17c-06) Recombinant Monoclonal Antibody for Western Blot
The Human Parechovirus Type 1/3 VP3 protein is a structural component of the human parechoviruses (HPeVs), specifically serotypes 1 and 3, which are members of the Picornaviridae family. The VP3 protein is a part of the viral capsid, which encapsulates the viral RNA genome, providing protection and facilitating the virus's entry into host cells. Human parechoviruses are known to cause a range of illnesses, from mild gastrointestinal and respiratory infections to more severe conditions such as meningitis, encephalitis, and neonatal sepsis, particularly in infants and young children. The VP3 protein, along with other capsid proteins (VP1 and VP2), plays a crucial role in the virus's ability to infect host cells and elicit an immune response. Due to its essential functions in the viral life cycle and its immunogenic properties, VP3 is a significant target for diagnostic assays, vaccine development, and antiviral research. Understanding the structure and function of the VP3 protein in HPeV-1 and HPeV-3 is vital for developing effective strategies to prevent and treat infections caused by these viruses.
Specifications
| Human Parechovirus 1/3 VP3 (Strain: WA17c-06) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2443 | |
| Unconjugated | |
| Human PeV 1/3 VP3 | |
| Full length human Parechovirus 3 VP3 recombinant protein. (Strain#WA17c-06). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction