missing translation for 'onlineSavingsMsg'
Learn More

Human Pappalysin-2/PAPP-A2 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18793857
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793857 25 μg 25µg
18169120 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18793857 Supplier R&D Systems Supplier No. MAB1668SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pappalysin-2/PAPP-A2 Monoclonal specifically detects Pappalysin-2/PAPP-A2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pappalysin-2/PAPP-A2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 242011
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BXP8
Gene Alias EC 3.4.24.-, PAPPA2, PAPP-A2, PAPP-A2pappalysin-2, pappalysin 2, Pappalysin2, PAPPE, PAPP-E, PLAC3, placenta-specific 3, pregnancy-associated plasma preproprotein-A2, Pregnancy-associated plasma protein A2, Pregnancy-associated plasma protein E1
Gene Symbols PAPPA2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pappalysin-2/PAPP-A2 Ser234-Cys1396 Accession # Q9BXP8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60676
Test Specificity Detects human Pappalysin-2/PAPP-A2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human PAPP-A is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.