missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pappalysin-2/PAPP-A2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1668
This item is not returnable.
View return policy
Description
Pappalysin-2/PAPP-A2 Monoclonal specifically detects Pappalysin-2/PAPP-A2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Pappalysin-2/PAPP-A2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9BXP8 | |
| PAPPA2 | |
| Mouse myeloma cell line NS0-derived recombinant human Pappalysin-2/PAPP-A2 Ser234-Cys1396 Accession # Q9BXP8 | |
| 500 μg | |
| Primary | |
| Detects human Pappalysin-2/PAPP-A2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human PAPP-A is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 242011 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24.-, PAPPA2, PAPP-A2, PAPP-A2pappalysin-2, pappalysin 2, Pappalysin2, PAPPE, PAPP-E, PLAC3, placenta-specific 3, pregnancy-associated plasma preproprotein-A2, Pregnancy-associated plasma protein A2, Pregnancy-associated plasma protein E1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 60676 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction