missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Pappalysin-1/PAPP-A DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
The Human Pappalysin-1/PAPP-A DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Pappalysin-1/PAPP-A.
Specifications
Specifications
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 0.78 to 50 ng/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 5069 |
| Synonym | ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4 ase, IGFBP-4 aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA, PAPPA1, PAPP-A1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, Pappalysin1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction