missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAPPA Recombinant Protein

Product Code. 11579011 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 11579011

Brand: Invitrogen™ RP75696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human Pregnancy Associated Plasma Protein-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13219
Concentration 3.0 mg/mL
For Use With (Application) Control, Western Blot
Formulation 20 mM tris HCl, 4 M urea with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 5069
Name Human PAPPA
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4 ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1
Common Name PAPPA
Gene Symbol PAPPA
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Arg81-Asp271 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.