missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PAPPA Recombinant Protein
Description
This product contains purified recombinant human Pregnancy Associated Plasma Protein-1 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).
Specifications
Specifications
| Accession Number | Q13219 |
| Concentration | 3.0 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | 20 mM tris HCl, 4 M urea with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 5069 |
| Name | Human PAPPA |
| Quantity | 10 μg |
| Source | E. coli |
| Immunogen | N/A |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?