missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pannexin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7088-SP
This item is not returnable.
View return policy
Description
Pannexin-2 Monoclonal specifically detects Pannexin-2 in Human samples. It is validated for Western Blot.
Specifications
| Pannexin-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q96RD6 | |
| PANX2 | |
| E. coli-derived recombinant human Pannexin-2 Arg316-Lys456 Accession # Q96RD6 | |
| 25 μg | |
| Primary | |
| Detects human Pannexin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) Pannexin-1 and no cross-reactivity with rhPannexin-3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 719406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hPANX2, MGC119432, pannexin 2, pannexin-2, Pannexin2, PANX2, PX2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56666 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction