missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Pancreatic Polypeptide/PP ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human Pancreatic Polypeptide/PP quantitates human Pancreatic Polypeptide/PP in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Pancreatic Polypeptide/PP.
Pancreatic polypeptide (PP) is a 36 amino-acid residue peptide with sequence and 3-D structural homology with NPY and PYY. PP is the least conserved of the three peptides and was the first to be identified. It is almost exclusively expressed in the endocrine pancreas and released in response to cholinergic stimulation and food intake. PP has inhibitory effects on gall bladder secretion, intestinal motility and pancreatic secretion. PP may serve as a marker for certain gut endocrine tumors including carcinoid tumors.
Specifications
Specifications
| Accession Number | AV061413; C-terminal peptide; HGNC:9327; IGSPP; IOPPP; Obinepitide; panceatic polypeptide; Pancreatic hormone; Pancreatic icosapeptide; Pancreatic polypeptide; pancreatic polypeptide Y; pancreatic prohormone; Pancreatic prohormone percursor; pancreatic prohormone precursor; PH; PI; PNP; PP; PP1; PPA1; PPase; Ppy; prepro-PP (prepropancreatic polypeptide); SID6-8061 |
| Assay Range | 4 to 1,000 pg/mL |
| Assay Sensitivity | 4 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5539 |
| Gene Symbol | PPY |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction