missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PAM Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4837
This item is not returnable.
View return policy
Description
Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Human samples. It is validated for Western Blot.
Specifications
| Peptidylglycine alpha-Amidating Monooxygenase/PAM | |
| Polyclonal | |
| Unconjugated | |
| P19021 | |
| PAM | |
| Mouse myeloma cell line NS0-derived recombinant human PAM Phe21-Val817 Accession # P19021 | |
| 100 μg | |
| Primary | |
| Detects human PAM in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5066 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido