missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PAK1IP1 (aa 301-365) Control Fragment Recombinant Protein

Product Code. 30195671
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195671

Brand: Invitrogen™ RP94639

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (51%), Rat (51%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56349 (PA5-56349. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Negatively regulates the PAK1 kinase. PAK1 is a member of the PAK kinase family, which has been shown to play a positive role in the regulation of signaling pathways involving MAPK8 and RELA. PAK1 exists as an inactive homodimer, which is activated by binding of small GTPases such as CDC42 to an N-terminal regulatory domain. PAK1IP1 also binds to the N-terminus of PAK1, and inhibits the specific activation of PAK1 by CDC42. May be involved in ribosomal large subunit assembly (PubMed:24120868). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NWT1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55003
Name Human PAK1IP1 (aa 301-365) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5830431I15Rik; 5930415H02Rik; AA419825; AI314040; AW556169; bA421M1.5; Gdpd1; glycerophosphodiester phosphodiesterase domain containing 1; hPIP1; MAK11; p21-activated protein kinase-interacting protein 1; PAK/PLC-interacting protein 1; PAK1 interacting protein 1; PAK1-interacting protein 1; PAK1IP1; PIP1; putative PAK inhibitor Skb15; RP11-421M1.5; WD repeat-containing protein 84; WDR84
Common Name PAK1IP1
Gene Symbol PAK1IP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VWLDKVADMKESLPPAAEPSPVSKEQSKIGKKEPGDTVHKEEKRSKPNTKKRGLTGDSKKATKES
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.