missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human PAF-AH Recombinant Protein, PeproTech®

Product Code. 17881343
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17881343

Brand: Gibco™ 14010100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human PAF-AH is a 420 amino acid glycoprotein which migrates with an apparent molecular mass of 47-55 kDa by SDS-PAGE analysis. Recombinan Human PAF-AH has a calculated, theoretical molecular weight of 47.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Platelet Activating Factor (PAF) is a biologically active phospholipid, which exerts primarily proinflammatory activities by specifically signaling through G-protein-coupled receptors on platelets, neutrophils, and monocytes. Platelet Activating Factor Acetylhydrolase (PAF-AH) is a secreted protein that mediates PAF activity by specifically catalyzing hydrolysis of the 'sn2' ester bond, resulting in the conversion of PAF to the biologically inactive lyso-PAF. PAF-AH can also interact with LDL particles to induce the hydrolysis of LDL-associated, oxidized phospholipids, generating lysophosphatidylcholine (lyso-PC) and other lysophospholipids.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13093
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7941
Molecular Weight (g/mol) 47.8 kDa
Name Human PAF-AH
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1-alkyl-2-acetylglycerophosphocholine esterase; 2-acetyl-1-alkylglycerophosphocholine esterase; Group-VIIA phospholipase A2; gVIIA-PLA2; LDL-associated phospholipase A2; LDL-PLA(2); LDL-PLA2; lipoprotein-associated phospholipase A2; LP-PLA2; PAF 2-acylhydrolase; PAF acetylhydrolase; PAFAD; Pafah; phospholipase A2 group VII; phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma); PLA2G7; Platelet-activating factor acetylhydrolase; R75400
Common Name PLA2G7
Gene Symbol PLA2G7
Biological Activity Measured by its ability to cleave a PAF analog in a chromogenic substrate linked assay. At a PAF-AH concentration of 10.0 ug/ml, 50% cleavage was achieved (at an incubation time of approximately 2 minutes.)
Conjugate Unconjugated
Recombinant Recombinant
Sequence FDWQYINPVA HMKSSAWVNK IQVLMAAASF GQTKIPRGNG PYSVGCTDLM FDHTNKGTFL RLYYPSQDND RLDTLWIPNK EYFWGLSKFL GTHWLMGNIL RLLFGSMTTP ANWNSPLRPG EKYPLVVFSH GLGAFRTLYS AIGIDLASHG FIVAAVEHRD RSASATYYFK DQSAAEIGDK SWLYLRTLKQ EEETHIRNEQ VRQRAKECSQ ALSLILDIDH GKPVKNALDL KFDMEQLKDS IDREKIAVIG HSFGGATVIQ TLSEDQRFRC GIALDAWMFP LGDEVYSRIP QPLFFINSEY FQYPANIIKM KKCYSPDKER KMITIRGSVH QNFADFTFAT GKIIGHMLKL KGDIDSNVAI DLSNKASLAF LQKHLGLHKD FDQWDCLIEG DDENLIPGTN INTTNQHIML QNSSGIEKYN
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.