missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PACT Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1980-SP
This item is not returnable.
View return policy
Description
PACT Polyclonal specifically detects PACT in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| PACT | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DYT16, DYT16interferon-inducible double stranded RNA-dependent activator, HSD14, PACTPKR-associating protein X, PKR-associated protein X, PKR-Associating Protein X, PRKRA, protein kinase, interferon-inducible double stranded RNA dependent activator, RAX, RAXHSD14 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8575 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| O75569 | |
| PRKRA | |
| E. coli-derived recombinant human PACT Ser2-Lys313 Accession # O75569 | |
| 25 μg | |
| Primary | |
| Detects human PACT in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction