missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p70 S6 Kinase beta Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2987
This item is not returnable.
View return policy
Description
p70 S6 Kinase beta/S6K2 Polyclonal specifically detects p70 S6 Kinase beta/S6K2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| p70 S6 Kinase beta/S6K2 | |
| Polyclonal | |
| Unconjugated | |
| Q9UBS0 | |
| RPS6KB2 | |
| E. coli-derived recombinant human p70 S6 Kinase beta Ala2-Ser410 Accession # Q9UBS0 | |
| 100 μg | |
| Primary | |
| Detects human p70 S6 Kinase beta in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human p70 S6 Kinase is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, RPS6KB2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14B, STK14BS6 kinase-related kinase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6199 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction