missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human p67phox Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7830-SP
This item is not returnable.
View return policy
Description
p67phox/NOXA2 Polyclonal specifically detects p67phox/NOXA2 in Human samples. It is validated for Western Blot.
Specifications
| p67phox/NOXA2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| autosomal 2, NADPH oxidase activator 2, NCF2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2, NOXA2FLJ93058, p67phox, p67-phox | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4688 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P19878 | |
| NCF2 | |
| E. coli-derived recombinant human p67phox Lys355-Val526 (His389Gln) Accession # P19878 | |
| 25 μg | |
| Primary | |
| Detects human p67phox in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction