missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human p53R2 Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0095

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15256319

  • £112.00 / 50µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

p53R2 Synthetic Peptide, PEP-0095, from Invitrogen.

This peptide corresponds to 16 amino amino terminus of human p53R2.PEP-0095 can be used as a blocking peptide with polyclonal antibody PA5-19970.
Specifications

Specifications

50484
purified
Human p53R2
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q7LG56
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
RRM2B
0.02% sodium azide
50 μg
PBS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers