missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human p400 (aa 946-1086) Control Fragment Recombinant Protein

Product Code. 30210748
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210748

Brand: Invitrogen™ RP91695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53475 (PA5-53475. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. May be required for transcriptional activation of E2F1 and MYC target genes during cellular proliferation. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. May regulate ZNF42 transcription activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96L91
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57634
Name Human p400 (aa 946-1086) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700020J09Rik; AU023439; CAG repeat protein 32; CAGH32; Domino homolog; E1A binding protein p400; E1A-binding protein p400; EP400; hDomino; KIAA1498; KIAA1818; mDomino; mKIAA1498; P400; p400 kDa SWI2/SNF2-related protein; p400 SWI2/SNF2-related protein; TNRC12; trinucleotide repeat containing 12; trinucleotide repeat-containing gene 12 protein
Common Name p400
Gene Symbol EP400
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DDEVDDEEETIEEEEANEGVVDHQTELSNLAKEAELPLLDLMKLYEGAFLPSSQWPRPKPDGEDTSGEEDADDCPGDRESRKDLVLIDSLFIMDQFKAAERMNIGKPNAKDIADVTAVAEAILPKGSARVTTSVKFNAPSL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.